Learn More
Bochem™ 18/10 Stainless Steel Lab Jacks
Brand:  Bochem™ 11132
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10088521
  • 2677.50 SEK / Each
Estimated Shipment: 21-12-2021
Specifications
Stainless Steel
200mm
200 L x 200 Wmm
18/10 Stainless Steel
Wheels
Lab Jack
200mm
1Each