Learn More
Bochem™ 18/10 Stainless Steel Paddle Stirrers with 3 Holes
356.00 SEK - 368.00 SEK
Specifications
Diameter (Metric) | 70 mm |
---|---|
Diameter (Metric) Shaft | 8 mm |
Includes | 3 Holes |
Quantity | 1 Each |
Type | Paddle Stirrer |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10060891
|
Bochem™
9405 |
500 mm |
368.00 SEK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
10490153
|
Bochem™
9404 |
400 mm |
356.00 SEK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Specifications
70 mm | |
3 Holes | |
Paddle Stirrer |
8 mm | |
1 Each |