missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Vibro Spatulas

Brand:  Bochem™ 3540

Additional Details : Weight : 0.10000kg

Product Code. 10516742

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

1 Ea.
50 mm
12 mm
190 x 12 mm
Stainless Steel (18/10)
Vibro Spatula
12 mm
Stainless Steel (18/10)
Plastic
190 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers