Läs mer
Spare Pyrex™ stopcock glass keys
Material:
Glas
Förpackningsstorlek:
Varje
Beskrivning
- Replacement keys for SSD: series stopcocks
- Manufactured from borosilicate glass
- Supplied complete with key retaining device
Specifikationer
Specifikationer
| Material | Glas |
| Produkttyp | Glass key and retaining set |