Läs mer
18-Oxocortisol, TRC
Produktkod. 30008385
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008385 5 mg
30006352 10 mg
30007814 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008385 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8706155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2410-60-8
Molekylformel C21H28O6
Molekylvikt (g/mol) 376.44
Synonym 11β,17,21-Trihydroxy-3,20-dioxo-pregn-4-en-18-al, (11β)-11,17,21-trihydroxy-3,20-diox-pregn-4-en-18-al
LEDER >>[H][C@]12[C@@]([C@@](CC[C@@]3(C(CO)=O)O)([H])[C@]3(C=O)C[C@@H]2O)([H])CCC4=CC(CC[C@@]41C)=O
Specifikationer
Kemiskt namn eller material 18-Oxocortisol
Kvantitet 5 mg
InChI formel InChI=1S/C21H28O6/c1-19-6-4-13(24)8-12(19)2-3-14-15-5-7-21(27,17(26)10-22)20(15,11-23)9-16(25)18(14)19/h8,11,14-16,18,22,25,27H,2-7,9-10H2,1H3/t14-,15-,16-,18+,19-,20+,21-/m0/s1