missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals MagFlex-96 aNAP System (RUO)

Product Code. 18954601 Shop All MP Biomedicals Products
Change view
Click to view available options
Capacity:
96 Samples/Run
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
18954601 96 Samples/Run Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18954601 Supplier MP Biomedicals Supplier No. 116013500
I lager
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fully automated high-throughput platform for magnetic bead-based nucleic acid purification.

  • The MagFlex-96 aNAP System is capable of processing up to 96 samples per run, it delivers high-quality DNA or RNA from a wide range of sample types, including blood, tissues, cells, swabs, plants, microbes, and environmental samples
  • With user-friendly preset programs and rapid processing, the system ensures consistent results and minimal hands-on time

Specifications

Product Type Nucleic Acid Extraction System
Sample Type Tissue, Blood, Bacteria, Fresh or Frozen Plant Tissue, Tissue Homogenate
Suitable Downstream Analysis Next-Generation Sequencing
Volume (Metric) Processing 50 to 1000 μL
Plates per Deck 6
Format 96-well Plate
Capacity 96 Samples/Run
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.