Läs mer
18Alpha-Glycyrrhizic Acid Ammonium Salt ~85%, TRC
Produktkod. 30133582
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133582 5 mg
30137994 50 mg
30012770 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133582 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG7351655MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80287-34-9
Molekylformel C42H62O16 . NH3
Molekylvikt (g/mol) 840
Synonym 18alphaH-Glycyrrhizic Acid Ammonium Salt, 18alphaH-Glycyrrhizinic Acid Ammonium Salt, Yiganmei Ammonium Salt, alpha-Glycyrrhizin Ammonium Salt, 18alpha-Glycyrrhizin, 18alpha-Glycyrrhizinic Acid Ammonium Salt, alpha-Glycyrrhizin Ammonium Salt, (3beta,18alpha,20beta)-20-Carboxy-11-oxo-30-norolean-12-en-3-yl 2-O-beta-D-Glucopyranuronosyl-alpha-D-glucopyranosiduronic Acid Ammonium Salt;
IUPAC-namn (2S,3S,4S,5R,6R)-6-[(2S,3R,4S,5S,6S)-2-[[(3S,4aR,6aR,6bS,8aS,11S,12aS,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-2,3,4a,5,6,7,8,9,10,12,12a,14a-dodecahydro-1H-picen-3-yl]oxy]-6-carboxy-4,5-dihydroxyoxan-3-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid;azane
LEDER N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]3(C)[C@@H]2C(=O)C=C4[C@H]5C[C@](C)(CC[C@]5(C)CC[C@@]34C)C(=O)O)O[C@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O[C@H]7O[C@H]([C@H](O)[C@@H](O)[C@@H]7O)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material 18alpha-Glycyrrhizic Acid Ammonium Salt
Kvantitet 5 mg
InChI formel InChI=1S/C42H62O16.H3N/c1-37(2)21-8-11-42(7)31(20(43)16-18-19-17-39(4,36(53)54)13-12-38(19,3)14-15-41(18,42)6)40(21,5)10-9-22(37)55-35-30(26(47)25(46)29(57-35)33(51)52)58-34-27(48)23(44)24(45)28(56-34)32(49)50;/h16,19,21-31,34-35,44-48H,8-15,17H2,1-7H3,(H,49,50)(H,51,52)(H,53,54);1H3/t19-,21+,22+,23+,24+,25+,26+,27-,28+,29+,30-,31-,34+,35+,38-,39+,40+,41-,42-;/m1./s1
Formel vikt 839.4303