Läs mer
1alpha-(Chloromethyl) Chlormadinone Acetate, TRC
Produktkod. 30134354
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134354 5 g
30138525 1 g
30006042 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134354 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3690855G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17183-98-1
Molekylformel C24 H30 Cl2 O4
Molekylvikt (g/mol) 453.4
Synonym Pregna-4,6-diene-3,20-dione, 17-(acetyloxy)-6-chloro-1-(chloromethyl)-, (1α)- (9CI), Pregna-4,6-diene-3,20-dione, 6-chloro-1α-(chloromethyl)-17-hydroxy-, acetate (8CI), 1α-(Chloromethyl) Chlormadinone Acetate, 6-Chloro-1α-(chloromethyl)-3,20-dioxopregna-4,6-dien-17-yl Acetate, 6-Chloro-1α-(chloromethyl)-17α-hydroxypregna-4,6-diene-3,20-dione 17-Acetate
IUPAC-namn [(1S,8R,9S,10S,13S,14S,17R)-17-acetyl-6-chloro-1-(chloromethyl)-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
LEDER CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)C[C@H](CCl)[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
Specifikationer
Kemiskt namn eller material 1alpha-(Chloromethyl) Chlormadinone acetate
Kvantitet 5 g
InChI formel InChI=1S/C24H30Cl2O4/c1-13(27)24(30-14(2)28)8-6-18-17-11-21(26)20-10-16(29)9-15(12-25)23(20,4)19(17)5-7-22(18,24)3/h10-11,15,17-19H,5-9,12H2,1-4H3/t15-,17+,18+,19+,22+,23-,24+/m1/s1
Formel vikt 452.1521