Läs mer
1H-1,2,3-Triazole-1-acetic Acid, TRC
Produktkod. 30138708
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138708 50 mg
17910515 5 mg
17921775 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138708 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT76754550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4314-22-1
Molekylformel C4H5N3O2
Molekylvikt (g/mol) 127.1
IUPAC-namn 2-(triazol-1-yl)acetic acid
LEDER OC(=O)Cn1ccnn1
Specifikationer
Kemiskt namn eller material 1H-1,2,3-Triazole-1-acetic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C4H5N3O2/c8-4(9)3-7-2-1-5-6-7/h1-2H,3H2,(H,8,9)
Formel vikt 127.0382