missing translation for 'onlineSavingsMsg'
Läs mer

1H-1-Ethyl-d5 Candesartan Cilexetil, TRC

Produktkod. 30373033
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373033 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30373033 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9008725MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 1H-1-Ethyl-d5 Candesartan Cilexetil
CAS 1246819-47-5
Molekylformel C35H33D5N6O6
Kvantitet 5 mg
Synonym 2-Ethoxy-1-[[2'-(1-(ethyl-d5)-1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic Acid 1-[[(Cyclohexyloxy)carbonyl]oxy]ethyl Ester
InChI formel InChI=1S/C35H38N6O6/c1-4-41-32(37-38-39-41)28-15-10-9-14-27(28)25-20-18-24(19-21-25)22-40-31-29(16-11-17-30(31)36-34(40)44-5-2)33(42)45-23(3)46-35(43)47-26-12-7-6-8-13-26/h9-11,14-21,23,26H,4-8,12-13,22H2,1-3H3/i1D3,4D2
LEDER [2H]C([2H])([2H])C([2H])([2H])n1nnnc1c2ccccc2c3ccc(Cn4c(OCC)nc5cccc(C(=O)OC(C)OC(=O)OC6CCCCC6)c45)cc3
Molekylvikt (g/mol) 643.74
Formel vikt 643.3167
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.