Läs mer
1H,1H,2H,2H,3H,3H-Tridecafluoro-1-nonanol, TRC
Produktkod. 30456830
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30456830 5 g
1 options
Produktkod. 30456830 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7750835G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1H,1H,2H,2H,3H,3H-Tridecafluoro-1-nonanol
CAS 80806-68-4
Anteckningar om renhetsgrad GC
Molekylformel C9H7OF13
Kvantitet 5 g
Synonym 1H,1H,2H,2H,3H,3H-Perfluorononanol, 1-Nonanol','4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluoro- (9CI','ACI), 4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluoro-1-nonanol (ACI), 3-(Perfluorohexyl)propanol, 4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluoronon-1-ol, 4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluorononan-1-ol
InChI formel InChI=1S/C9H7F13O/c10-4(11,2-1-3-23)5(12,13)6(14,15)7(16,17)8(18,19)9(20,21)22/h23H,1-3H2
LEDER OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluorononan-1-ol
Molekylvikt (g/mol) 378.13
Formel vikt 378.0289