Läs mer
1H,1H,2H,2H-Perfluorodecyl Acetate, TRC
Produktkod. 30419677
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419677 250 mg
30411793 100 mg
30410970 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30419677 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP286480250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 37858-04-1
Molekylformel C12 H7 F17 O2
Molekylvikt (g/mol) 506.1556
Synonym 1H,1H,2H,2H-Perfluorodecyl acetate, 1-Decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, 1-acetate, 1-Decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, acetate (9CI), 1,1,2,2-Tetrahydroperfluorodecyl acetate
IUPAC-namn 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl acetate
LEDER CC(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material 1H,1H,2H,2H-Perfluorodecyl Acetate
Kvantitet 250 mg
InChI formel InChI=1S/C12H7F17O2/c1-4(30)31-3-2-5(13,14)6(15,16)7(17,18)8(19,20)9(21,22)10(23,24)11(25,26)12(27,28)29/h2-3H2,1H3
Formel vikt 506.0175