missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1H,1H,2H,2H-Perfluorodecyltriethoxysilane is used to bond poly(tetrafluoroethylene) films to silicon wafers. It is also utilized in anti-reflective coating, release coating and soil repellent coating.
Solubility
Miscible with ethanol and tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents.
1H,1H,2H,2H-Perfluorodecyltriethoxysilane is used to bond poly(tetrafluoroethylene) films to silicon wafers. It is also utilized in anti-reflective coating, release coating and soil repellent coating.
Solubility
Miscible with ethanol and tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents.
Specifica
Boiling Point | 103°C to 106°C (3mmHg) |
Quantity | 5 g |
Linear Formula | CF3(CF2)7(CH2)2Si(OCH2CH3)3 |
UN Number | UN1760 |
Flash Point | 100°C (212°F) |
Solubility Information | Miscible with ethanol and tetrahydrofuran. |
Formula Weight | 610.38 |
Percent Purity | 97% |
Refractive Index | 1.342 |
Sensitivity | Moisture sensitive |
Vedi altri risultati |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'