Läs mer
1H,1H,6H-Decafluorohexan-1-ol, TRC
Produktkod. 30353207
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353207 250 mg
30360685 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353207 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD210625250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1H,1H,6H-Decafluorohexan-1-ol
CAS 60838-60-0
Molekylformel C6H4F10O
Kvantitet 250 mg
Synonym 2,2,3,3,4,4,5,5,6,6-Decafluorohexan-1-ol
InChI formel InChI=1S/C6H4F10O/c7-2(8)4(11,12)6(15,16)5(13,14)3(9,10)1-17/h2,17H,1H2
LEDER OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F
IUPAC-namn 2,2,3,3,4,4,5,5,6,6-decafluorohexan-1-ol
Molekylvikt (g/mol) 282.08
Formel vikt 282.0102