Läs mer
1H,1H-Perfluorohexan-1-ol, TRC
Produktkod. 30428711
Change view
Kvantitet:
2.5 g
5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30428711 5 g -
30410863 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30428711 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2863505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1H,1H-Perfluorohexanol
CAS 423-46-1
Anteckningar om renhetsgrad GC
Molekylformel C6 H3 F11 O
Kvantitet 5 g
Synonym 2,2,3,3,4,4,5,5,6,6,6-Undecafluoro-1-hexanol, 1,1-Dihydroperfluorohexanol, 1H,1H-Perfluoro-1-hexanol
InChI formel InChI=1S/C6H3F11O/c7-2(8,1-18)3(9,10)4(11,12)5(13,14)6(15,16)17/h18H,1H2
LEDER OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexan-1-ol
Molekylvikt (g/mol) 300.07
Formel vikt 300.0008