missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ CD56 Monoclonal Antibody (MEM-188), FITC
GREENER_CHOICE

Produktkod. 11559132
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 Tests
Förpackningsstorlek:
100 tester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11559132 100 Tests 100 tester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11559132 Leverantör Invitrogen™ Leverantörsnummer MA119574
  • ONLINE EXCLUSIVE PRICE
    4630.00 SEK
    3935.50 SEK / 100 tester
    Save 694.50 SEK
    15% Av
 Begränsad artikel
Beräknad leverans: 31-08-2026
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

This antibody reacts with an extracellular epitope on a 180 kDa isoform of CD56 (NCAM) expressed in leukocytes. It has been suggested that the antibody MEM-188 could react with rhesus monkey lymphocytes. Reactivity with other NCAM isoforms has not been tested.

CD56, also known as neural cell adhesion molecule (NCAM), is a highly glycosylated transmembrane glycoprotein of the immunoglobulin family. It plays a crucial role in cell adhesion, migration, axonal growth, pathfinding, and synaptic plasticity. CD56 is ubiquitously expressed in the nervous system in isoforms ranging from 120-180 kDa and is involved in homotypic adhesion of neural cells. It mediates interactions by binding extracellular matrix components such as laminin and integrins, with polysialic modification reducing CD56-mediated adhesion. In the hematopoietic system, CD56 is expressed on natural killer (NK) cells and a subset of T cells known as NKT cells. It is also found on most neuroectodermal-derived cell lines, tissues, and neoplasms, including retinoblastoma, medulloblastoma, astrocytomas, and neuroblastoma. CD56 serves as a widely used neuroendocrine marker with high sensitivity for neuroendocrine tumors and ovarian granulosa cell tumors. Diseases associated with CD56 dysfunction include rabies and blastic plasmacytoid dendritic cell neoplasms, highlighting its importance in both neural and immune system functions.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen CD56
Användningsområden Flow Cytometry
Klassificering Monoclonal
Klona MEM-188
Koncentration 0.04 mg/mL
Konjugera FITC
Formulering PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gen Ncam1
Genaccessionsnr. P13591
Gene Alias adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM
Gensymboler Ncam1
Värd art Mouse
Immunogen KG-1 human acute myelogenous leukaemia cell line.
Reningsmetod Size-exclusion chromatography
Kvantitet 100 Tests
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 4684
Målarter Human, Monkey
Innehåll och lagring 4°C, store in dark, DO NOT FREEZE!
Produkttyp Antibody
Form Liquid
Isotyp IgG2a
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.