Learn More
1H-Indazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog Number p-4228737
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
61700-61-6
C8H6N2O2
162.15
MFCD06804570
MAVGBUDLHOOROM-UHFFFAOYSA-N
indazole-5-carboxylic acid, 5-indazolecarboxylic acid, 1h-indazole-5-carboxylicacid, indazole-5-carboxylic acid hcl, acmc-209mvj, 5-carboxy-1h-indazole, 5-carboxyindazole, 1h-5indazolecarboxylic acid, c8h6n2o2.clh, 1h-indazole-5carboxylic acid
4248454
1H-indazole-5-carboxylic acid
OC(=O)C1=CC=C2NN=CC2=C1
61700-61-6
C8H6N2O2
162.15
MFCD06804570
MAVGBUDLHOOROM-UHFFFAOYSA-N
indazole-5-carboxylic acid, 5-indazolecarboxylic acid, 1h-indazole-5-carboxylicacid, indazole-5-carboxylic acid hcl, acmc-209mvj, 5-carboxy-1h-indazole, 5-carboxyindazole, 1h-5indazolecarboxylic acid, c8h6n2o2.clh, 1h-indazole-5carboxylic acid
4248454
1H-indazole-5-carboxylic acid
OC(=O)C1=CC=C2NN=CC2=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 321.0°C to 324.0°C
Color Yellow to Amber
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 162.15
Percent Purity 97%
Physical Form Beads
Chemical Name or Material 1H-Indazole-5-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1H-Indazole-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.