Läs mer
1H-Pyrrolo[3,2-b]pyridine-2,3-dione, TRC
Produktkod. 30420815
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420815 100 mg
30412271 50 mg
30412712 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420815 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP903720100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 153072-89-0
Molekylformel C7H4N2O2
Molekylvikt (g/mol) 148.12
IUPAC-namn 1H-pyrrolo[3,2-b]pyridine-2,3-dione
LEDER O=C1Nc2cccnc2C1=O
Specifikationer
Kemiskt namn eller material 1H-Pyrrolo[3,2-b]pyridine-2,3-dione
Kvantitet 100 mg
InChI formel InChI=1S/C7H4N2O2/c10-6-5-4(9-7(6)11)2-1-3-8-5/h1-3H,(H,9,10,11)
Formel vikt 148.0273