Läs mer
Tocris Bioscience™ AP 811
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
Beskrivning
AP 811 is a high affinity, selective ANP clearance receptor (NPR3) antagonist (Ki = 0.45 nM). Exhibits >20,000-fold selectivity for NPR3 over NPR1. Inhibits ANP-stimulated Na+/K+ pump activity in rabbit ventricular cardiomyocytes in vitro. Also blocks proliferation of rodent neonatal cardiomyocytes seen at low ANP concentrations in vitro.
Specifikationer
Specifikationer
| Beskrivning | AP 811 |
| Molekylvikt (g/mol) | 915.1 |
| Produkttyp | AP 811 |
| Kvantitet | 1 mg |
| Sekvens | N2-[4-(2-Naphthoylamino)phenylacetyl]-Arg-Ile-Asp-Arg-(S)-2-methylbutylamide |