missing translation for 'onlineSavingsMsg'
Learn More

Carbon Steel, AISI 1050, G10500, ARMI∣MBH

Product Code. 17987384
missing translation for 'orderingAttributeHoverText'
Kvantitet:
1 Ea.
This item is not returnable. View return policy

Product Code. 17987384

missing translation for 'mfr': ARMI/MBH IARMFE105018G

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Carbon Steel, AISI 1050, G10500, Ø38x19mm
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Boron/Titanium/Niobium/Antimony/Oxygen/Aluminium/Arsenic/Zinc/Phosphorus/Cobalt/Nitrogen/Tin/Molybdenum/Sulfur/Vanadium/Nickel/Chromium/Copper/Silicon/Carbon/Manganese/Iron/Hydrogen/Cadmium/Bismuth/Magnesium/Selenium/Lead/Tungsten/Zirconium/Calcium/Tantalum
CAS 7439-89-6
Kvantitet 1 Ea.
Form Disc
Koncentration eller sammansättning (efter analyt eller komponenter) Boron 0.0005 %/Titanium 0.0008 %/Niobium 0.0013 %/Antimony 0.0015 %/Oxygen 0.0026 %/Aluminium 0.003 %/Arsenic 0.003 %/Zinc 0.003 %/Phosphorus 0.0045 %/Cobalt 0.0056 %/Nitrogen 0.0097 %/Tin 0.0103 %/Molybdenum 0.018 %/Sulfur 0.027 %/Vanadium 0.027 %/Nickel 0.068 %/Chromium 0.1 %/Copper 0.179 %/Silicon 0.223 %/Carbon 0.499 %/Manganese 0.79 %/Iron 98 %/Hydrogen (<0.0001) %/Cadmium (<0.0010) %/Bismuth (<0.0050) %/Magnesium (<0.0050) %/Selenium (<0.0050) %/Lead (0.00015) %/Tungsten (0.0005) %/Zirconium (0.001) %/Calcium (0.002) %/Tantalum (0.002) %
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.