Läs mer
(1R)-1-[3-[(E)-2-(7-Chloro-2-quinolyl)vinyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propan-1-ol, Mikromol™
Produktkod. 30051357
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051357 100 mg
30049307 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051357 Leverantör Mikromol Leverantörsnummer MM0698.21
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1R)-1-[3-[(E)-2-(7-Chloro-2-quinolyl)vinyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propan-1-ol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R)-1-(3-((E)-2-(7-Chloro-2-quinolyl)vinyl)phenyl)-3-(2-(1-hydroxy-1-methylethyl)phenyl)propan-1-ol
CAS 150026-75-8
Analyt- eller komponentnamn (1R)-1-(3-((E)-2-(7-Chloro-2-quinolyl)vinyl)phenyl)-3-(2-(1-hydroxy-1-methylethyl)phenyl)propan-1-ol
Frakt skick Room Temperature
Molekylformel C29 H28 Cl N O2
Certifikat och efterlevnad ISO/IEC 17025
Synonym α-[3-[(1E)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-2-[(1R)-1-hydroxy-1-methylethyl]benzenepropanol, Benzenepropanol, α-[3-[2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-2-(1-hydroxy-1-methylethyl)-, [R-(E)]- (9CI)
InChI formel InChI=1S/C29H28ClNO2/c1-29(2,33)26-9-4-3-7-21(26)13-17-28(32)23-8-5-6-20(18-23)10-15-25-16-12-22-11-14-24(30)19-27(22)31-25/h3-12,14-16,18-19,28,32-33H,13,17H2,1-2H3/b15-10+/t28-/m1/s1
LEDER CC(C)(O)c1ccccc1CC[C@@H](O)c2cccc(\C=C\c3ccc4ccc(Cl)cc4n3)c2
IUPAC-namn (1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propan-1-ol
Molekylvikt (g/mol) 457.99
Kvantitet 100 mg
Formel vikt 457.1809 g/mol