Läs mer
[1R-[1Alpha(R*),3aBeta,7aAlpha]]-Octahydro-Delta,7a-dimethyl-4-oxo-1H-indene-1-pentanoic Acid Methyl Ester, TRC
Produktkod. 30413371
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30413371 10 mg
1 options
Produktkod. 30413371 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO23645010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material [1R-(1alpha(R*),3abeta,7aalpha)]-Octahydro-delta,7a-dimethyl-4-oxo-1H-indene-1-pentanoic Acid Methyl Ester
CAS 135359-41-0
Molekylformel C17H28O3
Kvantitet 10 mg
Synonym [1R-[1A(R*),3Ab,7Aa]]-Octahydro-D,7A-Dimethyl-4-Oxo-1H-Indene-1-Pentanoic Acid Methyl Ester
InChI formel InChI=1S/C17H28O3/c1-12(6-4-8-16(19)20-3)13-9-10-14-15(18)7-5-11-17(13,14)2/h12-14H,4-11H2,1-3H3/t12-,13-,14+,17-/m1/s1
LEDER COC(=O)CCC[C@@H](C)[C@H]1CC[C@H]2C(=O)CCC[C@]12C
IUPAC-namn methyl (5R)-5-[(1R,3aR,7aR)-7a-methyl-4-oxo-2,3,3a,5,6,7-hexahydro-1H-inden-1-yl]hexanoate
Molekylvikt (g/mol) 280.4
Formel vikt 280.2038