Läs mer
(1R)-2,3-Dihydro-N-2-propen-1-yl-1H-inden-1-amine Ethanedioate, TRC
Produktkod. 30377353
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30377353 10 mg
1 options
Produktkod. 30377353 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45043510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R)-2,3-Dihydro-N-2-propen-1-yl-1H-inden-1-amine Ethanedioate
CAS 1166392-43-3
Molekylformel C21 H23 N O10
Kvantitet 10 mg
Synonym (R)-N-Allyl-2,3-dihydro-1H-inden-1-amine Oxalate, (R)-N-Allyl-1-aminoindane Oxalate, Rasagiline Impurity Oxalate Salt
InChI formel InChI=1S/C21H23NO10/c1-10(23)29-9-15-17(30-11(2)24)18(31-12(3)25)16(21(28-4)32-15)22-19(26)13-7-5-6-8-14(13)20(22)27/h5-8,15-18,21H,9H2,1-4H3/t15?,16?,17-,18?,21+/m0/s1
LEDER CO[C@@H]1OC(COC(=O)C)[C@H](OC(=O)C)C(OC(=O)C)C1N2C(=O)c3ccccc3C2=O
IUPAC-namn [(3R,6R)-3,4-diacetyloxy-5-(1,3-dioxoisoindol-2-yl)-6-methoxyoxan-2-yl]methyl acetate
Molekylvikt (g/mol) 449.408
Formel vikt 449.132