Läs mer
(1R,2R)-1,2-Cyclohexanedimethanol 1,2-Dimethanesulfonate, TRC
Produktkod. 30365887
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365887 25 mg
30366172 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30365887 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98795025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 186204-35-3
Molekylformel C10H20O6S2
Molekylvikt (g/mol) 300.39
Synonym (1R-trans)-1,2-Cyclohexanedimethanol Dimethanesulfonate, (1R,2R)-1,2-Bis(methanesulfonyloxymethyl)cyclohexane, (1R,2R)-1,2-Bis(methanesulfonyloxymethyl)cyclohexan
IUPAC-namn [(1R,2R)-2-(methylsulfonyloxymethyl)cyclohexyl]methyl methanesulfonate
LEDER CS(=O)(=O)OC[C@@H]1CCCC[C@H]1COS(=O)(=O)C
Specifikationer
Kemiskt namn eller material (1R,2R)-1,2-Cyclohexanedimethanol 1,2-Dimethanesulfonate
Kvantitet 25 mg
InChI formel InChI=1S/C10H20O6S2/c1-17(11,12)15-7-9-5-3-4-6-10(9)8-16-18(2,13)14/h9-10H,3-8H2,1-2H3/t9-,10-/m0/s1
Formel vikt 300.0701