Läs mer
(1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride, TRC
Produktkod. 30341379
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30341379 100 mg
1 options
Produktkod. 30341379 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA938150100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride
CAS 158414-44-9
Molekylformel C6H11NO2 . HCl
Kvantitet 100 mg
Synonym Cyclopentanecarboxylic acid, 2-amino-, hydrochloride, (1R-trans)- (9CI), (1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride, (1R,2R)-2-aminocyclopentanecarboxylic acid hydrochloride (1:1), (1R,2R)-2-Aminocyclopentane-1-carboxylic acid hydrochloride, trans-(1R,2R)-2-Aminocyclopentane-1-carboxylic acid hydrochloride, trans-(1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride
InChI formel InChI=1S/C6H11NO2.ClH/c7-5-3-1-2-4(5)6(8)9;/h4-5H,1-3,7H2,(H,8,9);1H/t4-,5-;/m1./s1
LEDER Cl.N[C@@H]1CCC[C@H]1C(=O)O
IUPAC-namn (1R,2R)-2-aminocyclopentane-1-carboxylic acid;hydrochloride
Molekylvikt (g/mol) 165.62
Formel vikt 165.0557