Läs mer
(1R,2R,3S,5R)-Pinanediol Pyrrolidine-2S-boronate Hydrochloride, TRC
Produktkod. 30426728
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30426728 25 mg
1 options
Produktkod. 30426728 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP45860525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R,2R,3S,5R)-Pinanediol Pyrrolidine-2S-boronate Hydrochloride
CAS 149716-73-4
Molekylformel C14H25BClNO2
Kvantitet 25 mg
Synonym (2S)-2-[(3aR,4R,6R,7aS)-hexahydro-3a,5,5-trimethyl-4,6-methano-1,3,2-benzodioxaborol-2-yl]pyrrolidine Hhydrochloride, [3aR-[2(S*),3aalpha,4beta,6beta,7aalpha]]-2-(Hexahydro-3a,5,5-trimethyl-4,6-methano-1,3,2-benzodioxaborol-2-yl)pyrrolidine Hydrochloride
InChI formel InChI=1S/C14H24BNO2.ClH/c1-13(2)9-7-10(13)14(3)11(8-9)17-15(18-14)12-5-4-6-16-12;/h9-12,16H,4-8H2,1-3H3;1H/t9-,10-,11+,12-,14-;/m1./s1
LEDER Cl.CC1(C)[C@H]2C[C@@H]3OB(O[C@]3(C)[C@@H]1C2)[C@H]4CCCN4
Molekylvikt (g/mol) 285.62