Läs mer
(1R,2R)-Bortezomib, TRC
Produktkod. 30012178
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012178 5 mg
30008298 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012178 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6757055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1132709-15-9
Molekylformel C19 H25 B N4 O4
Molekylvikt (g/mol) 384.24
Synonym Boronic acid, B-[(1R)-3-methyl-1-[[(2R)-1-oxo-3-phenyl-2-[(2-pyrazinylcarbonyl)amino]propyl]amino]butyl]-, B-[(1R)-3-Methyl-1-[[(2R)-1-oxo-3-phenyl-2-[(2-pyrazinylcarbonyl)amino]propyl]amino]butyl]boronic acid, bortezomib diastereomer (R,R) (USP), bortezomib related compound A (USP)
IUPAC-namn [(1R)-3-methyl-1-[[(2R)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoyl]amino]butyl]boronic acid
LEDER CC(C)C[C@H](NC(=O)[C@@H](Cc1ccccc1)NC(=O)c2cnccn2)B(O)O
Specifikationer
Kemiskt namn eller material (1R,2R)-Bortezomib
InChI formel InChI=1S/C19H25BN4O4/c1-13(2)10-17(20(27)28)24-18(25)15(11-14-6-4-3-5-7-14)23-19(26)16-12-21-8-9-22-16/h3-9,12-13,15,17,27-28H,10-11H2,1-2H3,(H,23,26)(H,24,25)/t15-,17+/m1/s1
Kvantitet 5 mg
Formel vikt 384.1969