missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1R,2R)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine, 97%

Brand:  Alfa Aesar™ H63540.ME

Additional Details : CAS Number : 174758-63-5 Weight : 0.00050kg

Quantity 500mg

 View more versions of this product

Product Code. 15468356

  • 2063.28 SEK / 500mg


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

(1R,2R)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine
174758-63-5
662.78
MFCD16618374

Specifications

(1R,2R)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine
>300°C
174758-63-5
Solid
662.78
Yellow
500mg
Air Sensitive
97%
MFCD16618374
Videos
SDS