Learn More
Alfa Aesar™ (1R,2R)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine, 97%
Brand:  Alfa Aesar™ H63540.MC
Additional Details : CAS Number : 174758-63-5 Weight : 0.00010kg
Quantity 100mg
 View more versions of this product
Product Code. 15458356
  • 754.93 SEK / 100mg
Estimated Shipment date
from Supplier 04-07-2018
Add to basket
Specifications
Chemical Identifiers
(1R,2R)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine
174758-63-5
662.78
MFCD16618374
Specifications
(1R,2R)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine
>300°C
174758-63-5
Solid
662.78
Yellow
100mg
Air Sensitive
97%
MFCD16618374