Läs mer
(1R,2S,5R)-(-)-Menthol Beta-D-Glucuronide, TRC
Produktkod. 30137745
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137745 100 mg
30004501 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137745 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM218880100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79466-08-3
Molekylformel C16H28O7
Molekylvikt (g/mol) 332.39
Synonym (1R,2S,5R)-(-)-Menthol Ss-D-Glucuronide
IUPAC-namn (2S,3S,6R)-3,4,5-trihydroxy-6-[(1R,2R,5R)-5-methyl-2-propan-2-ylcyclohexyl]oxyoxane-2-carboxylic acid
LEDER CC(C)[C@H]1CC[C@@H](C)C[C@H]1O[C@@H]2O[C@@H]([C@@H](O)C(O)C2O)C(=O)O
Specifikationer
Kemiskt namn eller material (1R,2S,5R)-(-)-Menthol beta-D-Glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C16H28O7/c1-7(2)9-5-4-8(3)6-10(9)22-16-13(19)11(17)12(18)14(23-16)15(20)21/h7-14,16-19H,4-6H2,1-3H3,(H,20,21)/t8-,9-,10-,11?,12+,13?,14+,16-/m1/s1
Formel vikt 332.1835
Procent renhet >95%