Läs mer
(1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine Acetate, Mikromol™
Beskrivning
(1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine Acetate
Specifikationer
Specifikationer
| Kemiskt namn eller material | (1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine Acetate |
| CAS | 2724762-78-9 |
| Analyt- eller komponentnamn | (1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine Acetate |
| Frakt skick | Room Temperature |
| Molekylformel | C9 H18 N2 . C2 H4 O2 |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | (3-endo)-9-Methyl-9-Azabicyclo[3.3.1]nonan-3-amine acetate (1:1), (1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine Acetate, Granisetron Hydrochloride Imp. E (EP) as Acetate, Granisetron Hydrochloride Related Compound E (USP) as Acetate Salt, Granisetron Related Compound E as Acetate, Granisetron Hydrochloride Impurity E as Acetate, Granisetron Impurity E as Acetate |
| InChI formel | InChI=1S/C9H18N2.C2H4O2/c1-11-8-3-2-4-9(11)6-7(10)5-8;1-2(3)4/h7-9H,2-6,10H2,1H3;1H3,(H,3,4)/t7-,8+,9-; |
| LEDER | CN1[C@@H]2CCC[C@H]1C[C@@H](N)C2.CC(=O)O |
| IUPAC-namn | acetic acid;(1R,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-amine |
| Visa mer |