Läs mer
1R,3R,AlphaR-Deltamethrin, TRC
Produktkod. 30012362
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012362 10 mg
30003293 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012362 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23076010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55700-99-7
Molekylformel C22H19Br2NO3
Molekylvikt (g/mol) 505.2
Synonym [1R-[1α(R*),3α]]-3-(2,2-Dibromoethenyl)-2,2-dimethyl-cyclopropanecarboxylic Acid Cyano(3-phenoxyphenyl)methyl Ester, NRDC 156A, RU 23938
LEDER CC1(C)[C@@H](C=C(Br)Br)[C@H]1C(=O)O[C@@H](C#N)c2cccc(Oc3ccccc3)c2
Specifikationer
Kemiskt namn eller material 1R,3R,aR-Deltamethrin
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C22H19Br2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17-,18-,20-/m0/s1
Formel vikt 502.973
Procent renhet >95%