Läs mer
(1R,3S)-(-)-3-Aminocyclopentanecarboxylic acid, 95%, Thermo Scientific™
Produktkod. p-7034437
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
Produktkod. 15409605
Brand: Thermo Scientific Alfa Aesar H52122.MD
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
(1R,3S)-(-)-3-Aminocyclopentanecarboxylic acid is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 71830-08-5
Molekylformel C6H11NO2
Molekylvikt (g/mol) 129.159
MDL-nummer MFCD00211289
InChI-nyckel MLLSSTJTARJLHK-UHNVWZDZSA-N
Synonym 1r,3s-3-aminocyclopentanecarboxylic acid, 1r,3s-3-aminocyclopentane-1-carboxylic acid, --1r,3s-3-aminocyclopentanecarboxylic acid, cis-3-aminocyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-amino-, 1r,3s, 1r,3s---3-aminocyclopentanecarboxylic acid, cis-3-amino-cyclopentanecarboxylic acid, 1r,3s-3-amino-cyclopentanecarboxylic acid, pubchem18731, 1r,3s---3-amino-cyclopentanecarboxylic acid
PubChem CID 1502035
IUPAC-namn (1R,3S)-3-aminocyclopentane-1-carboxylic acid
LEDER C1CC(CC1C(=O)O)N
Specifikationer
Kemiskt namn eller material (1R,3S)-(-)-3-Aminocyclopentanecarboxylic acid
Löslighetsinformation Slightly soluble in water.
Kvantitet 250 mg
Formel vikt 129.16
Känslighet Air Sensitive
Procent renhet 95%