Läs mer
(1R,3S)-3-Aminocyclopentanol Hydrochloride, TRC
Produktkod. 30327082
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30327082 100 mg
30327365 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30327082 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA596540100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R,3S)-3-Aminocyclopentanol Hydrochloride
CAS 1279032-31-3
Molekylformel C5 H11 N O . Cl H
Kvantitet 100 mg
Synonym (1R,3S)-3-Aminocyclopentan-1-ol Hydrochloride
InChI formel InChI=1S/C5H11NO.ClH/c6-4-1-2-5(7)3-4;/h4-5,7H,1-3,6H2;1H/t4-,5+;/m0./s1
LEDER Cl.N[C@H]1CC[C@@H](O)C1
IUPAC-namn (1R,3S)-3-aminocyclopentanol;hydrochloride
Molekylvikt (g/mol) 137.61
Formel vikt 137.0607