Läs mer
(1R,3S,5S)-3-(2-Hydroxy-2,2-di(thiophen-2-yl)acetoxy)-8,8-dimethyl-8-azabicyclo[3.2.1]oct-6-en-8-ium Bromide, TRC
Produktkod. 30138640
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138640 250 mg
30135300 25 mg
30007266 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH941715250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136310-95-7
Molekylformel C19 H22 N O3 S2 . Br
Molekylvikt (g/mol) 456.42
Synonym 8-Azoniabicyclo[3.2.1]oct-6-ene, 3-[(hydroxydi-2-thienylacetyl)oxy]-8,8-dimethyl-, bromide, (3-endo)- (9CI), 8-Azoniabicyclo[3.2.1]oct-6-ene, 3-[(hydroxydi-2-thienylacetyl)oxy]-8,8-dimethyl-, bromide, endo-, 8-Azoniabicyclo[3.2.1]oct-6-ene, 3-[(2-hydroxy-2,2-di-2-thienylacetyl)oxy]-8,8-dimethyl-, bromide (1:1), (3-endo)-, Tiotropium Bromide Monohydrate Imp C (EP)
IUPAC-namn [(1S,5R)-8,8-dimethyl-8-azoniabicyclo[3.2.1]oct-6-en-3-yl] 2-hydroxy-2,2-dithiophen-2-ylacetate;bromide
LEDER [Br-].C[N+]1(C)[C@@H]2C[C@H](C[C@H]1C=C2)OC(=O)C(O)(c3cccs3)c4cccs4
Specifikationer
Kemiskt namn eller material (1R,3S,5S)-3-(2-Hydroxy-2,2-di(thiophen-2-yl)acetoxy)-8,8-dimethyl-8-azabicyclo[3.2.1]oct-6-en-8-ium Bromide
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C19H22NO3S2.BrH/c1-20(2)13-7-8-14(20)12-15(11-13)23-18(21)19(22,16-5-3-9-24-16)17-6-4-10-25-17;/h3-10,13-15,22H,11-12H2,1-2H3;1H/q+1;/p-1/t13-,14+,15+;
Formel vikt 455.0224
Procent renhet >95%