Läs mer
(1R,3S)-Solifenacin Succinate, TRC
Produktkod. 30426821
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30426821 25 mg -
30417472 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30426821 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS67669525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1262506-09-1
Molekylformel C23H26N2O2 . C4H6O4
Molekylvikt (g/mol) 480.6
Synonym Butanedioic acid compd. with (3S)-1-azabicyclo[2.2.2]oct-3-yl (1R)-3,4-dihydro-1-phenyl-2(1H)-isoquinolinecarboxylate, (3S)-1-Azabicyclo[2.2.2]oct-3-yl (1R)-3,4-dihydro-1-phenyl-2(1H)-isoquinolinecarboxylate butanedioate
IUPAC-namn [(3S)-1-azabicyclo[2.2.2]octan-3-yl] (1R)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate;butanedioic acid
LEDER OC(=O)CCC(=O)O.O=C(O[C@@H]1CN2CCC1CC2)N3CCc4ccccc4[C@H]3c5ccccc5
Specifikationer
Kemiskt namn eller material (1R,3S)-Solifenacin Succinate
Kvantitet 25 mg
InChI formel InChI=1S/C23H26N2O2.C4H6O4/c26-23(27-21-16-24-13-10-18(21)11-14-24)25-15-12-17-6-4-5-9-20(17)22(25)19-7-2-1-3-8-19;5-3(6)1-2-4(7)8/h1-9,18,21-22H,10-16H2;1-2H2,(H,5,6)(H,7,8)/t21-,22-;/m1./s1
Formel vikt 480.226