Läs mer
(1R-4S)-N-[4-Chloro-5-(formylamino)-6-[[4-(hydroxymethyl)-2-cyclopenten-1-yl]amino]-2-pyrimidinyl]acetamide, TRC
Produktkod. 30368756
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368756 500 mg
30358747 100 mg
30356641 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30368756 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC366495500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136522-32-2
Molekylformel C13H16ClN5O3
Molekylvikt (g/mol) 325.75
Synonym Acetamide, N-[4-chloro-5-(formylamino)-6-[[4-(hydroxymethyl)-2-cyclopenten-1-yl]amino]-2-pyrimidinyl]-, (1R-cis)-, N-[4-Chloro-5-formamido-6-[[(1R,4S)-4-(hydroxymethyl)cyclopent-2-en-1-yl]amino]pyrimidin-2-yl]acetamide
IUPAC-namn N-[4-chloro-5-formamido-6-[[(1R,4S)-4-(hydroxymethyl)cyclopent-2-en-1-yl]amino]pyrimidin-2-yl]acetamide
LEDER CC(=O)Nc1nc(Cl)c(NC=O)c(N[C@@H]2C[C@H](CO)C=C2)n1
Specifikationer
Kemiskt namn eller material (1R-4S)-N-(4-Chloro-5-(formylamino)-6-((4-(hydroxymethyl)-2-cyclopenten-1-yl)amino)-2-pyrimidinyl)acetamide
Kvantitet 500 mg
InChI formel InChI=1S/C13H16ClN5O3/c1-7(22)16-13-18-11(14)10(15-6-21)12(19-13)17-9-3-2-8(4-9)5-20/h2-3,6,8-9,20H,4-5H2,1H3,(H,15,21)(H2,16,17,18,19,22)/t8-,9+/m1/s1
Formel vikt 325.0942