Läs mer
(1R,4S)-rel-4-(Acetylamino)-2-cyclopentene-1-carboxylic Acid Methyl Ester, TRC
Produktkod. 30328055
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30328055 100 mg
1 options
Produktkod. 30328055 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA168245100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R,4S)-rel-4-(Acetylamino)-2-cyclopentene-1-carboxylic Acid Methyl Ester
CAS 61865-49-4
Molekylformel C9H13NO3
Kvantitet 100 mg
Synonym cis-(+/-)-4-(Acetylamino)-2-Cyclopentene-1-carboxylic Acid Methyl Ester
InChI formel InChI=1S/C9H13NO3/c1-6(11)10-8-4-3-7(5-8)9(12)13-2/h3-4,7-8H,5H2,1-2H3,(H,10,11)/t7-,8+/m1/s1
LEDER COC(=O)[C@H]1C[C@@H](NC(=O)C)C=C1
IUPAC-namn methyl (1S,4R)-4-acetamidocyclopent-2-ene-1-carboxylate
Molekylvikt (g/mol) 183.2
Formel vikt 183.0895