Läs mer
(1R,4S)-Sertraline Hydrochloride, TRC
Produktkod. 30419425
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30419425 1 mg
1 options
Produktkod. 30419425 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2800101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R,4S) Sertraline Hydrochloride
CAS 79896-32-5
Molekylformel C17H18Cl3N
Kvantitet 1 mg
Synonym (1R,4S)-Sertraline HCl
InChI formel InChI=1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17-;/m0./s1
LEDER Cl.CN[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13
Molekylvikt (g/mol) 342.69