Läs mer
(1R,5R,6R)-5-(1-Ethylpropoxy)-7-oxabicyclo[4.1.0]hept-3-ene-3-carboxylic Acid Ethyl Ester, TRC
Produktkod. 30374211
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374211 50 mg
30379881 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30374211 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92569050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1266663-89-1
Molekylformel C14H22O4
Molekylvikt (g/mol) 254.32
Synonym Oseltamivir Impurity 3
IUPAC-namn ethyl (1R,5R,6R)-5-(pentan-3-yloxy)-7-oxabicyclo[4.1.0]hept-3-ene-3-carboxylate
LEDER O=C(OCC)C1=C[C@@H](OC(CC)CC)[C@H](O2)[C@H]2C1
Specifikationer
Kemiskt namn eller material (1R,5R,6R)-5-(1-Ethylpropoxy)-7-oxabicyclo[4.1.0]hept-3-ene-3-carboxylic Acid Ethyl Ester
Kvantitet 50 mg
InChI formel InChI=1S/C14H22O4/c1-4-10(5-2)17-11-7-9(14(15)16-6-3)8-12-13(11)18-12/h7,10-13H,4-6,8H2,1-3H3/t11-,12-,13+/m1/s1
Formel vikt 254.15