Läs mer
(1R,5R,6R)-rel-5-(1-Ethylpropoxy)-7-oxabicyclo[4.1.0]hept-3-ene-3-carboxylic Acid Ethyl Ester, TRC
Produktkod. 30378355
Change view
Kvantitet:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378355 25 mg
30371669 10 mg
30377130 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30378355 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92569525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1217834-81-5
Molekylformel C14H22O4
Molekylvikt (g/mol) 254.32
LEDER O=C(OCC)C1=C[C@H](OC(CC)CC)[C@H](O2)[C@H]2C1
Specifikationer
Kemiskt namn eller material (1R,5R,6R)-rel-5-(1-Ethylpropoxy)-7-oxabicyclo[4.1.0]hept-3-ene-3-carboxylic Acid Ethyl Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C14H22O4/c1-4-10(5-2)17-11-7-9(14(15)16-6-3)8-12-13(11)18-12/h7,10-13H,4-6,8H2,1-3H3/t11-,12+,13-/m0/s1
Formel vikt 254.1518
Procent renhet >95%