Läs mer
[1R-(exo,exo)]-8-Methyl-3-phenyl-8-azabicyclo[3.2.1]octane-2-carboxylic Acid Methyl Ester, TRC
Produktkod. 30472478
Change view
Kvantitet:
5 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472478 25 mg
30475388 5 mg
30469326 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30472478 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33111025MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50372-80-0
Molekylformel C16H21NO2
Molekylvikt (g/mol) 259.34
Synonym Win 33065-2, Win 35065-2, (1R,2S,3S,5S)-8-Methyl-3-phenyl-8-azabicyclo[3.2.1]octane-2-carboxylic Acid Methyl Ester
IUPAC-namn methyl (1S,3S,4S,5R)-8-methyl-3-phenyl-8-azabicyclo[3.2.1]octane-4-carboxylate
LEDER COC(=O)[C@@H]1[C@H]2CC[C@@H](C[C@@H]1c3ccccc3)N2C
Specifikationer
Kemiskt namn eller material [1R-(exo,exo)]-8-Methyl-3-phenyl-8-azabicyclo[3.2.1]octane-2-carboxylic Acid Methyl Ester
Kvantitet 25 mg
InChI formel InChI=1S/C16H21NO2/c1-17-12-8-9-14(17)15(16(18)19-2)13(10-12)11-6-4-3-5-7-11/h3-7,12-15H,8-10H2,1-2H3/t12-,13+,14+,15-/m0/s1
Formel vikt 259.1572