Läs mer
(-)-(1R)-Menthyl Chloroformate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (-)-(1R)-Menthyl Chloroformate |
| CAS | 14602-86-9 |
| Molekylformel | C11H19ClO2 |
| Kvantitet | 1 g |
| Synonym | Carbonochloridic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester, (-)-(1R,2S,5R)-Menthyl Chloroformate, (-)-(R)-Menthyl Chloroformate, (-)-Menthyl Chloroformate, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Chloroformate, (1R,2S,5R)-Menthyl Chloroformate, Chlorocarbonic Acid (-)-Menthyl Ester, L-(-)-Menthyl Chloroformate, L-Menthoxy Chloroformate, L-Menthyl Chloroformate, Menthyl Chloroformate |
| InChI formel | InChI=1S/C11H19ClO2/c1-7(2)9-5-4-8(3)6-10(9)14-11(12)13/h7-10H,4-6H2,1-3H3 |
| LEDER | CO/N=C(/c1nc(NCOC(=O)C(C)(C)C)sc1)C(=O)N[C@H]2[C@H]3N(C(C(=O)OCOC(=O)C(C)(C)C)=C(\C=C/c4c(C)ncs4)CS3)C2=O |
| Molekylvikt (g/mol) | 218.72 |