Läs mer
(1RS)-1-[4-Hydroxy-3-(hydroxymethyl)phenyl]-2-[(4-phenylbutyl)amino]ethanol, Mikromol™
Produktkod. 17942216
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
17942216 25 mg
1 options
Produktkod. 17942216 Leverantör Mikromol Leverantörsnummer MM0536.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1RS)-1-[4-Hydroxy-3-(hydroxymethyl)phenyl]-2-[(4-phenylbutyl)amino]ethanol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1RS)-1-(4-Hydroxy-3-(hydroxymethyl)phenyl)-2-((4-phenylbutyl)amino)ethanol
CAS 1798014-51-3
Analyt- eller komponentnamn (1RS)-1-(4-Hydroxy-3-(hydroxymethyl)phenyl)-2-((4-phenylbutyl)amino)ethanol
Frakt skick Room Temperature
Molekylformel C19 H25 N O3
Certifikat och efterlevnad ISO 17025
Synonym Salbutamol Sulphate Imp. C (EP), Salbutamol Imp. C (EP), Albuterol USP Related Compound A, Albuterol USP RC A, Benzenemethanol, alpha-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-3-methyl-, Benzyl alcohol, alpha-[(tert-butylamino)methyl]-4-hydroxy-3-methyl- (8CI), alpha-[[(1,1-Dimethylethyl)amino]methyl]-4-hydroxy-3-methylbenzenemethanol, (1RS)-2-[(1,1-Dimethylethyl)amino]-1-(4-hydroxy-3-methylphenyl)ethanol, AH 4045X, USP Albuterol Related Compound A, Ph Eur Salbutamol Impurity C, (1RS)-1-[4-Hydroxy-3-(hydroxymethyl)phenyl]-2-[(4-phenylbutyl)amino]ethanol, Salmeterol Xinafoate Imp. A (EP), Salmeterol USP Related Compound A, Salmeterol USP RC A, (1RS)-1-[4-hydroxy-3-(hydroxymethyl)phenyl]-2-[(4-phenylbutyl)amino]ethanol, Salmeterol Related Compound A (USP), GR 73664X, USP Salmeterol Related Compound A, Ph Eur Salmeterol Impurity A, GR 73664X, Salmeterol Related Compound A, Salmeterol Xinafoate Impurity A, Salmeterol Impurity A
InChI formel InChI=1S/C19H25NO3/c21-14-17-12-16(9-10-18(17)22)19(23)13-20-11-5-4-8-15-6-2-1-3-7-15/h1-3,6-7,9-10,12,19-23H,4-5,8,11,13-14H2
LEDER OCc1cc(ccc1O)C(O)CNCCCCc2ccccc2
IUPAC-namn 2-(hydroxymethyl)-4-[1-hydroxy-2-(4-phenylbutylamino)ethyl]phenol
Molekylvikt (g/mol) 315.41
Kvantitet 25 mg
Formel vikt 315.1834 g/mol