Läs mer
(1RS)-2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanol, Mikromol™
Produktkod. 30050380
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050380 25 mg
30050203 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050380 Leverantör Mikromol Leverantörsnummer MM0381.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1RS)-2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1RS)-2-(Benzyl(1,1-dimethylethyl)amino)-1-(4-hydroxy-3-(hydroxymethyl)phenyl)ethanol
CAS 24085-03-8
Analyt- eller komponentnamn (1RS)-2-(Benzyl(1,1-dimethylethyl)amino)-1-(4-hydroxy-3-(hydroxymethyl)phenyl)ethanol
Frakt skick Room Temperature
Molekylformel C20 H27 N O3
Certifikat och efterlevnad ISO 17025
Synonym Salbutamol Sulphate Imp. E (EP), Salbutamol Imp. E (EP), (1RS)-2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanol, AH 3521, Ph Eur Salbutamol Impurity E, Benzyl salbutamol, Salbutamol Impurity E, Salbutamol Sulfate Impurity E, Salbutamol Impurity E
InChI formel InChI=1S/C20H27NO3/c1-20(2,3)21(12-15-7-5-4-6-8-15)13-19(24)16-9-10-18(23)17(11-16)14-22/h4-11,19,22-24H,12-14H2,1-3H3
LEDER CC(C)(C)N(CC(O)c1ccc(O)c(CO)c1)Cc2ccccc2
IUPAC-namn 4-[2-[benzyl(tert-butyl)amino]-1-hydroxyethyl]-2-(hydroxymethyl)phenol
Molekylvikt (g/mol) 329.43
Kvantitet 25 mg
Formel vikt 329.1991 g/mol