Läs mer
[(1S,2R)-1-Benzyl-2-hydroxy-3-[isobutyl-[(4-aminophenyl)sulfonyl]amino]propyl]carbamic Acid tert-Butyl Ester, TRC
Produktkod. 30339518
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30339518 250 mg
1 options
Produktkod. 30339518 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB279950250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material tert-Butyl N-[(1S,2R)-1-Benzyl-2-hydroxy-3-[(4-aminophenyl)sulfonyl(2-methylpropyl)amino]propyl]carbamate
CAS 183004-94-6
Molekylformel C25 H37 N3 O5 S
Kvantitet 250 mg
Synonym Carbamic acid, N-[(1S,2R)-3-[[(4-aminophenyl)sulfonyl](2-methylpropyl)amino]-2-hydroxy-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester, Carbamic acid, [(1S,2R)-3-[[(4-aminophenyl)sulfonyl](2-methylpropyl)amino]-2-hydroxy-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester (9CI), Carbamic acid, [3-[[(4-aminophenyl)sulfonyl](2-methylpropyl)amino]-2-hydroxy-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester, [R-(R*,S*)]-, tert-Butyl [(1S,2R)-3-[[(4-aminophenyl)sulfonyl](isobutyl)amino]-1-benzyl-2-hydroxypropyl]carbamate
InChI formel InChI=1S/C25H37N3O5S/c1-18(2)16-28(34(31,32)21-13-11-20(26)12-14-21)17-23(29)22(15-19-9-7-6-8-10-19)27-24(30)33-25(3,4)5/h6-14,18,22-23,29H,15-17,26H2,1-5H3,(H,27,30)/t22-,23+/m0/s1
LEDER CC(C)CN(C[C@@H](O)[C@H](Cc1ccccc1)NC(=O)OC(C)(C)C)S(=O)(=O)c2ccc(N)cc2
IUPAC-namn tert-butyl N-[(2S,3R)-4-[(4-aminophenyl)sulfonyl-(2-methylpropyl)amino]-3-hydroxy-1-phenylbutan-2-yl]carbamate
Molekylvikt (g/mol) 491.64
Formel vikt 491.2454