Läs mer
(1S,2R,5S)-(+)-Menthol, TRC
Produktkod. 30137995
Change view
Kvantitet:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137995 10 g
30011251 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137995 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM21888510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15356-60-2
Molekylformel C10 H20 O
Molekylvikt (g/mol) 156.27
Synonym Cyclohexanol, 5-methyl-2-(1-methylethyl)-, (1S,2R,5S)-, Cyclohexanol, 5-methyl-2-(1-methylethyl)-, [1S-(1α,2β,5α)]-, Menthol, (1S,3S,4R)-(+)- (8CI), (1S,2R,5S)-5-Methyl-2-(1-methylethyl)cyclohexanol, (+)-Menthol, (1S,2R,5S)-(+)-Menthol, (1S,2R,5S)-2-Isopropyl-5-methylcyclohexanol, (1S,2R,5S)-Menthol, D-Menthol, D-Menthol, d-Menthol
IUPAC-namn (1S,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
LEDER CC(C)[C@H]1CC[C@H](C)C[C@@H]1O
Specifikationer
Kemiskt namn eller material (+)-Menthol
Anteckningar om renhetsgrad GC
Kvantitet 10 g
InChI formel InChI=1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m0/s1
Formel vikt 156.1514
Procent renhet >95%