missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-4229193
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
1091606-67-5
C26H24NP
381.46
MFCD11045444
MIPGTOGPLXZBQX-QYRYXDJWNA-N
1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
46177587
(1S,2S)-2-diphenylphosphanyl-1,2-diphenylethanamine
N[C@H]([C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1091606-67-5
C26H24NP
381.46
MFCD11045444
MIPGTOGPLXZBQX-QYRYXDJWNA-N
1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
46177587
(1S,2S)-2-diphenylphosphanyl-1,2-diphenylethanamine
N[C@H]([C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 87.0°C to 93.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 381.45
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (1S, 2S)-2-(Diphenylphosphino)-1, 2-diphenylethylamine
Show More Show Less
compliance-icons
Product Identifier
  • (1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'