Läs mer
(1S,2S)-N-Acetylpseudoephedrine, Mikromol™
Produktkod. 30052115
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052115 100 mg
30050548 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052115 Leverantör Mikromol Leverantörsnummer MM2115.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1S,2S)-N-Acetylpseudoephedrine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1S,2S)-N-Acetylpseudoephedrine
CAS 84472-25-3
Analyt- eller komponentnamn (1S,2S)-N-Acetylpseudoephedrine
Frakt skick Room Temperature
Molekylformel C12 H17 N O2
Certifikat och efterlevnad ISO 17025
Synonym (1S,2S)-N-Acetylpseudoephedrine
InChI formel InChI=1S/C12H17NO2/c1-9(13(3)10(2)14)12(15)11-7-5-4-6-8-11/h4-9,12,15H,1-3H3/t9-,12+/m0/s1
LEDER C[C@@H]([C@@H](O)c1ccccc1)N(C)C(=O)C
IUPAC-namn N-[(1S,2S)-1-hydroxy-1-phenylpropan-2-yl]-N-methylacetamide
Molekylvikt (g/mol) 207.27
Kvantitet 100 mg
Formel vikt 207.1259 g/mol