Learn More
(1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
100 mg
500 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is employed as a intermediate for pharmaceutical and asymmetirc synthesis.
Solubility
Insoluble in water.
Notes
Air Sensitive, store away from air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is employed as a intermediate for pharmaceutical and asymmetirc synthesis.
Solubility
Insoluble in water.
Notes
Air Sensitive, store away from air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Color | Yellow |
Odor | Unpleasant |
Quantity | 100 mg |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 662.78 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | (1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine |